1-(1-(3-Chloro-4-(2-chloro-4-(trifluoromethoxy)phenoxy)pyridin-2-yl)piperidin-4-yl)-3-(pyridin-3-yl)thiourea 98%

Supplier: Ambeed
Warning

Synonyms: DO-264

A1001774-10MG A1001774-1MG A1001774-25MG A1001774-50MG A1001774-5MG
76670-736EA 100.79 USD
76670-736 76670-738 76670-740 76670-742 76670-744
1-(1-(3-Chloro-4-(2-chloro-4-(trifluoromethoxy)phenoxy)pyridin-2-yl)piperidin-4-yl)-3-(pyridin-3-yl)thiourea 98%
1-(1-(3-Chloro-4-(2-chloro-4-(trifluoromethoxy)phenoxy)pyridin-2-yl)piperidin-4-yl)-3-(pyridin-3-yl)thiourea

Precaución: for Research Use Only

Formula: C₂₃H₂₀Cl₂F₃N₅O₂S
MW: 558.4 g/mol
CAS Number: 2301866-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR