AZD 3241 98+%

Supplier: Ambeed
Warning

Synonyms: Verdiperstat, 1-(2-Isopropoxyethyl)-2-thioxo-2,3-dihydro-1H-pyrrolo[3,2-d]pyrimidin-4(5H)-one

A1001768-100MG A1001768-10MG A1001768-25MG A1001768-50MG A1001768-5MG A1001768-1MG
76670-726EA 514 USD
76670-726 76670-728 76670-730 76670-732 76670-734 77595-638
AZD 3241 98+%
AZD 3241

Precaución: for Research Use Only

Formula: C₁₁H₁₅N₃O₂S
MDL Number: MFCD30533444
CAS Number: 890655-80-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR