N1-(7-Chloroquinolin-4-yl)-N6-(6-((7-chloroquinolin-4-yl)amino)hexyl)-N6-methylhexane-1,6-diamine 98%

Supplier: Ambeed
Warning

A1001762-10MG A1001762-1MG A1001762-25MG A1001762-5MG
76670-712EA 75 USD
76670-712 76670-714 76670-716 76670-718
N1-(7-Chloroquinolin-4-yl)-N6-(6-((7-chloroquinolin-4-yl)amino)hexyl)-N6-methylhexane-1,6-diamine 98%
N1-(7-Chloroquinolin-4-yl)-N6-(6-((7-chloroquinolin-4-yl)amino)hexyl)-N6-methylhexane-1,6-diamine

Precaución: for Research Use Only

Formula: C₃₁H₃₉Cl₂N₅
MDL Number: MFCD31812586
CAS Number: 1872387-43-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR