4,4'-Bis(trifluoromethyl)-2,2'-bipyridyl 97%

Supplier: Ambeed
Warning

Synonyms: 4,4'-Bis(trifluoromethyl)-2,2'-bipyridine

A1001693-100MG A1001693-10G A1001693-1G A1001693-250MG A1001693-5G A1001693-25G
76670-702EA 10.61 USD
76670-702 76670-704 76670-706 76670-708 76670-710 77595-634
4,4'-Bis(trifluoromethyl)-2,2'-bipyridyl 97%
4,4'-Bis(trifluoromethyl)-2,2'-bipyridyl

Precaución: for Research Use Only

Formula: C₁₂H₆F₆N₂
MW: 292.18 g/mol
MDL Number: MFCD00233878
CAS Number: 142946-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR