1,4-Diiodo-2,3,5,6-tetramethylbenzene 95%

Supplier: Ambeed
Warning

Synonyms: 3,6-Diiododurene

A998010-100G A998010-1G A998010-25G A998010-5G
76669-816EA 495 USD
76669-816 76669-818 76669-820 76669-822
1,4-Diiodo-2,3,5,6-tetramethylbenzene 95%
1,4-Diiodo-2,3,5,6-tetramethylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂I₂
MW: 386.01 g/mol
MDL Number: MFCD00142554
CAS Number: 3268-21-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR