N-Methyl-7-nitro-1,2,3,4-tetrahydroquinoline 97%

Supplier: Ambeed
Warning

A665302-100MG A665302-1G A665302-250MG A665302-5G
76669-808EA 8 USD
76669-808 76669-810 76669-812 76669-814
N-Methyl-7-nitro-1,2,3,4-tetrahydroquinoline 97%
N-Methyl-7-nitro-1,2,3,4-tetrahydroquinoline

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₂O₂
MDL Number: MFCD00496655
CAS Number: 39275-18-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR