2-Fluoroadenosine 96%

Supplier: Ambeed
Warning

Synonyms: (2R,3R,4S,5R)-2-(6-Amino-2-fluoro-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol

A299140-10G A299140-1G
76669-776EA 265.23 USD
76669-776 76669-778
2-Fluoroadenosine 96%
2-Fluoroadenosine

Precaución: for Research Use Only

Formula: C₁₀H₁₂FN₅O₄
MW: 285.23 g/mol
MDL Number: MFCD00866394
CAS Number: 146-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR