Carbazochrome 97%

Supplier: Ambeed
Warning

A258034-10G A258034-1G A258034-250MG A258034-25G A258034-5G
76669-766EA 42 USD
76669-766 76669-768 76669-770 76669-772 76669-774
Carbazochrome 97%
Carbazochrome

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₄O₃
MDL Number: MFCD00059705
CAS Number: 69-81-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR