Ethylenediaminetetraacetic dianhydride 95%

Supplier: Ambeed
Warning

Synonyms: EDTA Dianhydride, 4,4'-Ethylenebis(2,6-morpholinedione), 4,4-(Ethane-1,2-diyl)bis(morpholine-2,6-dione)

A181777-100G A181777-1G A181777-25G A181777-500G A181777-5G
76669-750EA 80 USD
76669-750 76669-752 76669-754 76669-756 76669-758
Ethylenediaminetetraacetic dianhydride 95%
Ethylenediaminetetraacetic dianhydride

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₂O₆
MDL Number: MFCD00074963
CAS Number: 23911-25-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR