2-(4-Methylphenyl)propanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-(p-Tolyl)propanoic acid

A180130-100G A180130-25G
76669-748EA 38.2 USD
76669-748 77083-352
2-(4-Methylphenyl)propanoic acid 98%
2-(4-Methylphenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂
MW: 164.21 g/mol
MDL Number: MFCD01111378
CAS Number: 938-94-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR