2-(Cyclopentyloxy)-6-fluoropyridine 95+%

Supplier: Ambeed
Warning

A1683980-1G A1683980-250MG A1683980-5G
76669-692EA 118.83 USD
76669-692 76669-694 76669-696
2-(Cyclopentyloxy)-6-fluoropyridine 95+%
2-(Cyclopentyloxy)-6-fluoropyridine

Precaución: for Research Use Only

Formula: C₁₀H₁₂FNO
MW: 181.21 g/mol
MDL Number: MFCD14599963
CAS Number: 1249179-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR