Chloro(crotyl)(tri-tert-butylphosphine)palladium(II) 98+%

Supplier: Ambeed
Warning

Synonyms: PdCl(crotyl)(PtBu{3})

A1683678-100MG A1683678-1G A1683678-250MG
76669-678EA 93.36 USD
76669-678 77567-470 77567-472
Chloro(crotyl)(tri-tert-butylphosphine)palladium(II) 98+%
Chloro(crotyl)(tri-tert-butylphosphine)palladium(II)

Precaución: For research use only.

Formula: C₁₆H₃₅ClPPd
MW: 400.30 g/mol
MDL Number: MFCD25372547
CAS Number: 1334497-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR