1-(2-(Difluoromethyl)-5-methylphenyl)ethan-1-one 95%

Supplier: Ambeed
Warning

A1682917-5G A1682917-100MG A1682917-1G A1682917-250MG
76669-640EA 3362 USD
76669-640 77876-682 77876-683 77876-684
1-(2-(Difluoromethyl)-5-methylphenyl)ethan-1-one 95%
1-(2-(Difluoromethyl)-5-methylphenyl)ethan-1-one

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₂O
MDL Number: MFCD28521615
CAS Number: 1780446-98-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR