2-(3-(Butylsulfinyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
Supplier: Ambeed
|
Danger
|
A1681593-1G
A1681593-250MG
A1681593-5G
76669-614EA
209
USD
76669-614
76669-616
76669-618
2-(3-(Butylsulfinyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
2-(3-(Butylsulfinyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Precaución: for Research Use Only
Formula:
C₁₆H₂₅BO₃S MW: 308.24 g/mol |
UN:
2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...