4-Bromo-N-(4-fluoro-3-methoxyphenyl)benzenesulfonamide 98%

Supplier: Ambeed
Warning

A1681541-1G A1681541-250MG A1681541-5G
76669-608EA 209 USD
76669-608 76669-610 76669-612
4-Bromo-N-(4-fluoro-3-methoxyphenyl)benzenesulfonamide 98%
4-Bromo-N-(4-fluoro-3-methoxyphenyl)benzenesulfonamide

Precaución: for Research Use Only

Formula: C₁₃H₁₁BrFNO₃S
CAS Number: 1778962-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR