2-(3-(Butylsulfonyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%

Supplier: Ambeed
Warning

A1681529-1G A1681529-250MG A1681529-5G
76669-602EA 200.52 USD
76669-602 76669-604 76669-606
2-(3-(Butylsulfonyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
2-(3-(Butylsulfonyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₆H₂₅BO₄S
MW: 324.24 g/mol
CAS Number: 1801921-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR