(4-(3-Propylureido)phenyl)boronic acid 95%

Supplier: Ambeed
Warning

A1676797-100MG A1676797-1G A1676797-250MG
76669-470EA 47 USD
76669-470 76669-472 76669-474
(4-(3-Propylureido)phenyl)boronic acid 95%
(4-(3-Propylureido)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₅BN₂O₃
MW: 222.05 g/mol
MDL Number: MFCD31650940
CAS Number: 1360471-12-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR