(4-(3-Ethylureido)phenyl)boronic acid 95%

Supplier: Ambeed
Warning

A1676795-100MG A1676795-250MG A1676795-1G
76669-466EA 46 USD
76669-466 76669-468 77625-670
(4-(3-Ethylureido)phenyl)boronic acid 95%
(4-(3-Ethylureido)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₃BN₂O₃
MW: 208.02 g/mol
MDL Number: MFCD18383958
CAS Number: 1201597-61-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR