1-(2-Methoxyethyl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2(1H)-one 95+%

Supplier: Ambeed
Warning

A1672707-100MG A1672707-1G A1672707-250MG
76669-426EA 45.62 USD
76669-426 76669-428 76669-430
1-(2-Methoxyethyl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2(1H)-one 95+%
1-(2-Methoxyethyl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₁₄H₂₂BNO₄
MW: 279.14 g/mol
CAS Number: 1349734-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR