1-(Phenylsulfonyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole 95%

Supplier: Ambeed
Warning

A1672659-1G A1672659-250MG A1672659-5G
76669-414EA 219 USD
76669-414 76669-416 76669-418
1-(Phenylsulfonyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole 95%
1-(Phenylsulfonyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole

Precaución: for Research Use Only

Formula: C₂₀H₂₂BNO₄S
MDL Number: MFCD12828205
CAS Number: 1334950-73-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR