(4-((4-Chlorophenyl)ethynyl)phenyl)boronic acid 95%

Supplier: Ambeed
Warning

A1672429-100MG A1672429-1G A1672429-250MG
76669-402EA 45.62 USD
76669-402 76669-404 76669-406
(4-((4-Chlorophenyl)ethynyl)phenyl)boronic acid 95%
(4-((4-Chlorophenyl)ethynyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₀BClO₂
MW: 256.49 g/mol
CAS Number: 170629-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR