(1-((Tetrahydro-2H-pyran-4-yl)methyl)-1H-pyrazol-4-yl)boronic acid 95%
Supplier: Ambeed
|
Warning
|
A1672184-1G
A1672184-250MG
76669-386EA
323.58
USD
76669-386
76669-388
(1-((Tetrahydro-2H-pyran-4-yl)methyl)-1H-pyrazol-4-yl)boronic acid 95%
(1-((Tetrahydro-2H-pyran-4-yl)methyl)-1H-pyrazol-4-yl)boronic acid
Precaución: for Research Use Only
Formula:
C₉H₁₅BN₂O₃ MW: 210.03 g/mol |
MDL Number:
MFCD28969434 CAS Number: 2246772-81-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...