2-Bromo-4-fluoro-N,N-dimethylbenzenesulfonamide 95%

Supplier: Ambeed
Warning

A1672152-1G A1672152-250MG A1672152-25G A1672152-5G
76669-350EA 52 USD
76669-350 76669-352 76669-354 76669-356
2-Bromo-4-fluoro-N,N-dimethylbenzenesulfonamide 95%
2-Bromo-4-fluoro-N,N-dimethylbenzenesulfonamide

Precaución: for Research Use Only

Formula: C₈H₉BrFNO₂S
MW: 282.13 g/mol
MDL Number: MFCD12157938
CAS Number: 1183891-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR