N-Benzyl-2-bromo-4-methylbenzenesulfonamide 95%

Supplier: Ambeed
Warning

A1672150-1G A1672150-250MG A1672150-5G
76669-344EA 210.06 USD
76669-344 76669-346 76669-348
N-Benzyl-2-bromo-4-methylbenzenesulfonamide 95%
N-Benzyl-2-bromo-4-methylbenzenesulfonamide

Precaución: for Research Use Only

Formula: C₁₄H₁₄BrNO₂S
MW: 340.23 g/mol
MDL Number: MFCD12651434
CAS Number: 1184443-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR