1-(5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)piperazine x2,2,2-trifluoroacetate 95%

Supplier: Ambeed
Warning

A1670655-1G A1670655-250MG A1670655-5G
76669-326EA 49.87 USD
76669-326 76669-328 76669-330
1-(5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)piperazine x2,2,2-trifluoroacetate 95%
1-(5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)piperazine x2,2,2-trifluoroacetate

Precaución: for Research Use Only

Formula: C₁₇H₂₅BF₃N₃O₄
MW: 403.20 g/mol
CAS Number: 2782776-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR