Ethyl 4-iodo-1-(4-methoxybenzyl)-1H-pyrazole-5-carboxylate 95%

Supplier: Ambeed
Warning

A1669302-1G A1669302-100MG A1669302-250MG
76669-280EA 216 USD
76669-280 76768-884 76768-886
Ethyl 4-iodo-1-(4-methoxybenzyl)-1H-pyrazole-5-carboxylate 95%
Ethyl 4-iodo-1-(4-methoxybenzyl)-1H-pyrazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₅IN₂O₃
MDL Number: MFCD32666906
CAS Number: 1355336-09-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR