(R)-2-(4-(tert-Butyl)-4,5-dihydrooxazol-2-yl)phenol 95%

Supplier: Ambeed
Warning

A1665849-100MG A1665849-250MG
76669-222EA 212.18 USD
76669-222 77598-132
(R)-2-(4-(tert-Butyl)-4,5-dihydrooxazol-2-yl)phenol 95%
(R)-2-(4-(tert-Butyl)-4,5-dihydrooxazol-2-yl)phenol

Precaución: for Research Use Only

Formula: C₁₃H₁₇NO₂
MW: 219.28 g/mol
CAS Number: 150699-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR