2-(4-Chloro-3-(2-methoxyethoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%

Supplier: Ambeed
Warning

A1665845-1G A1665845-250MG A1665845-5G
76669-216EA 129.43 USD
76669-216 76669-218 76669-220
2-(4-Chloro-3-(2-methoxyethoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
2-(4-Chloro-3-(2-methoxyethoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₅H₂₂BClO₄
MW: 312.59 g/mol
CAS Number: 1853186-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR