N-(4-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)pivalamide 95%

Supplier: Ambeed
Warning

A1665531-100MG A1665531-250MG
76669-188EA 44.56 USD
76669-188 76669-190
N-(4-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)pivalamide 95%
N-(4-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)pivalamide

Precaución: for Research Use Only

Formula: C₁₈H₂₈BNO₃
MW: 317.23 g/mol
CAS Number: 2246585-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR