Methyl 5-methyl-2-morpholinothiazole-4-carboxylate 95%

Supplier: Ambeed
Warning

A1665077-100MG A1665077-250MG
76669-154EA 40 USD
76669-154 76669-156
Methyl 5-methyl-2-morpholinothiazole-4-carboxylate 95%
Methyl 5-methyl-2-morpholinothiazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₂O₃S
MDL Number: MFCD23652764
CAS Number: 1478718-44-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR