(4-((4-Acetylpiperazin-1-yl)methyl)phenyl)boronic acid 95%

Supplier: Ambeed
Warning

A1664921-1G A1664921-250MG
76669-150EA 140 USD
76669-150 76669-152
(4-((4-Acetylpiperazin-1-yl)methyl)phenyl)boronic acid 95%
(4-((4-Acetylpiperazin-1-yl)methyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₉BN₂O₃
CAS Number: 1082366-52-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR