Ethyl hept-6-ynoate 98%

Supplier: Ambeed
Warning

A1660686-100MG A1660686-1G A1660686-250MG
76669-138EA 65.78 USD
76669-138 76669-140 76669-142
Ethyl hept-6-ynoate 98%
Ethyl hept-6-ynoate

Precaución: for Research Use Only

Formula: C₉H₁₄O₂
MW: 154.21 g/mol
MDL Number: MFCD22041859
CAS Number: 105339-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR