4-Methoxy-3-(3-morpholinopropoxy)benzonitrile 97%

Supplier: Ambeed
Warning

A1638947-1G A1638947-5G
76669-128EA 65.78 USD
76669-128 76669-130
4-Methoxy-3-(3-morpholinopropoxy)benzonitrile 97%
4-Methoxy-3-(3-morpholinopropoxy)benzonitrile

Precaución: for Research Use Only

Formula: C₁₅H₂₀N₂O₃
MW: 276.33 g/mol
MDL Number: MFCD23134620
CAS Number: 675126-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR