2-Piperidino-5-pyrimidineboronic acid pinacol ester 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(Piperidin-1-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine, 2-(Piperiden-1-yl)pyrimidine-5-boronic acid, pinacol ester
A1638901-1G
A1638901-250MG
A1638901-5G
76669-122EA
101.85
USD
76669-122
76669-124
76669-126
2-Piperidino-5-pyrimidineboronic acid pinacol ester 98%
2-Piperidino-5-pyrimidineboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₅H₂₄BN₃O₂ MW: 289.1810 g/mol |
MDL Number:
MFCD07368250 CAS Number: 1015242-08-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...