2-Bromo-3,4-dichloro-1-methoxybenzene 95%

Supplier: Ambeed
Warning

A1636493-100MG A1636493-1G A1636493-250MG
76669-090EA 72 USD
76669-090 76669-092 76669-094
2-Bromo-3,4-dichloro-1-methoxybenzene 95%
2-Bromo-3,4-dichloro-1-methoxybenzene

Precaución: for Research Use Only

Formula: C₇H₅BrCl₂O
MDL Number: MFCD31725577
CAS Number: 174913-13-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR