1-(4-Methoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)ethan-1-one 97%

Supplier: Ambeed
Warning

A1636401-1G A1636401-5G
76669-086EA 25.47 USD
76669-086 76669-088
1-(4-Methoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)ethan-1-one 97%
1-(4-Methoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)ethan-1-one

Precaución: for Research Use Only

Formula: C₁₅H₂₁BO₄
MW: 276.13 g/mol
MDL Number: MFCD24039878
CAS Number: 1688672-91-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR