Ethyl 4-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 98%

Supplier: Ambeed
Warning

A1636206-1G A1636206-250MG A1636206-5G
76669-066EA 50 USD
76669-066 76669-068 76669-070
Ethyl 4-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 98%
Ethyl 4-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

Precaución: for Research Use Only

Formula: C₁₅H₂₀BFO₄
CAS Number: 474709-72-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR