3-Bromo-5-methyl-2-propoxypyridine 95%

Supplier: Ambeed
Warning

A1636046-1G A1636046-250MG A1636046-5G
76669-048EA 31.83 USD
76669-048 76669-050 76669-052
3-Bromo-5-methyl-2-propoxypyridine 95%
3-Bromo-5-methyl-2-propoxypyridine

Precaución: for Research Use Only

Formula: C₉H₁₂BrNO
MW: 230.10 g/mol
MDL Number: MFCD29114179
CAS Number: 1289096-91-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR