1-Chloro-3-fluoro-2-propoxybenzene 95%

Supplier: Ambeed
Warning

A1635764-1G A1635764-5G
76669-032EA 21 USD
76669-032 76669-034
1-Chloro-3-fluoro-2-propoxybenzene 95%
1-Chloro-3-fluoro-2-propoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₀ClFO
MW: 188.62 g/mol
CAS Number: 1865061-63-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR