2-(2,6-Difluoro-3-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%

Supplier: Ambeed
Warning

A1634516-1G A1634516-250MG A1634516-5G
76669-010EA 142 USD
76669-010 76669-012 76669-014
2-(2,6-Difluoro-3-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%
2-(2,6-Difluoro-3-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₃H₁₇BF₂O₂
MW: 254.08 g/mol
MDL Number: MFCD24310275
CAS Number: 2246738-41-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR