3-((2-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy)methyl)pyridine 95%

Supplier: Ambeed
Warning

A1631523-1G A1631523-250MG A1631523-5G
76668-992EA 78.51 USD
76668-992 76668-994 76668-996
3-((2-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy)methyl)pyridine 95%
3-((2-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy)methyl)pyridine

Precaución: for Research Use Only

Formula: C₁₈H₂₁BClNO₃
MW: 345.62 g/mol
CAS Number: 2828446-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR