(3-((2,5-Dioxopyrrolidin-1-yl)methyl)phenyl)boronic acid 95%

Supplier: Ambeed
Warning

A1631427-1G A1631427-250MG A1631427-5G
76668-980EA 161.26 USD
76668-980 76668-982 76668-984
(3-((2,5-Dioxopyrrolidin-1-yl)methyl)phenyl)boronic acid 95%
(3-((2,5-Dioxopyrrolidin-1-yl)methyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₂BNO₄
MW: 233.02 g/mol
MDL Number: MFCD28167438
CAS Number: 950576-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR