2-Isobutoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile 95%

Supplier: Ambeed
Warning

A1627908-1G A1627908-250MG A1627908-5G
76668-968EA 83 USD
76668-968 76668-970 76668-972
2-Isobutoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile 95%
2-Isobutoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₇H₂₄BNO₃
CAS Number: 2828446-41-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR