(7-Chloroquinolin-3-yl)boronic acid 95%

Supplier: Ambeed
Warning

A1627798-100MG A1627798-1G A1627798-250MG A1627798-5G
76668-950EA 248 USD
76668-950 76668-952 76668-954 76668-956
(7-Chloroquinolin-3-yl)boronic acid 95%
(7-Chloroquinolin-3-yl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₇BClNO₂
MDL Number: MFCD22394306
CAS Number: 1370040-83-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR