2-(3-((2-Chlorophenoxy)methyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%

Supplier: Ambeed
Warning

A1627412-1G A1627412-250MG A1627412-5G
76668-920EA 82.76 USD
76668-920 76668-922 76668-924
2-(3-((2-Chlorophenoxy)methyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
2-(3-((2-Chlorophenoxy)methyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₉H₂₂BClO₃
MW: 344.64 g/mol
CAS Number: 2828446-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR