2-(4-Chlorophenyl)benzo[d]thiazol-6-ol 95%

Supplier: Ambeed
Warning

A1627406-100MG A1627406-1G A1627406-250MG
76668-908EA 36.08 USD
76668-908 76668-910 76668-912
2-(4-Chlorophenyl)benzo[d]thiazol-6-ol 95%
2-(4-Chlorophenyl)benzo[d]thiazol-6-ol

Precaución: for Research Use Only

Formula: C₁₃H₈ClNOS
MW: 261.72 g/mol
MDL Number: MFCD19263714
CAS Number: 91692-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR