2-(3-Fluoro-4-(2,2,2-trifluoroethoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%

Supplier: Ambeed
Warning

A1626355-1G A1626355-250MG A1626355-5G
76668-896EA 117.76 USD
76668-896 76668-898 76668-900
2-(3-Fluoro-4-(2,2,2-trifluoroethoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%
2-(3-Fluoro-4-(2,2,2-trifluoroethoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₄H₁₇BF₄O₃
MW: 320.08 g/mol
MDL Number: MFCD32857797
CAS Number: 1689510-38-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR