2-(4-Fluorophenoxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 98%

Supplier: Ambeed
Warning

A1625452-1G A1625452-250MG A1625452-5G
76668-884EA 134.74 USD
76668-884 76668-886 76668-888
2-(4-Fluorophenoxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 98%
2-(4-Fluorophenoxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

Precaución: for Research Use Only

Formula: C₁₇H₁₉BFNO₃
MW: 315.14 g/mol
MDL Number: MFCD22683449
CAS Number: 1415825-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR