1-(3-(Benzyloxy)propyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 98%

Supplier: Ambeed
Warning

A1621744-1G A1621744-250MG A1621744-5G
76668-872EA 207.94 USD
76668-872 76668-874 76668-876
1-(3-(Benzyloxy)propyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 98%
1-(3-(Benzyloxy)propyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

Precaución: for Research Use Only

Formula: C₁₉H₂₇BN₂O₃
MW: 342.24 g/mol
MDL Number: MFCD22572272
CAS Number: 1309960-92-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR