3-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)benzoic acid 97%

Supplier: Ambeed
Warning

A1619989-1G A1619989-5G
76668-862EA 13.8 USD
76668-862 76668-864
3-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)benzoic acid 97%
3-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₅BO₄
MW: 234.05 g/mol
CAS Number: 1703741-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR